α,α,α,α',α',α'-Hexafluoro-2,5-xylidine
Leverancier: Molekula
MOLE30543599-1GEA
0
EUR
MOLE30543599-1G
α,α,α,α',α',α'-Hexafluoro-2,5-xylidine
α,α,α,α',α',α'-Hexafluoro-2,5-xylidine
Formule:
C₈H₅F₆N MW: 229,12 g/mol Kookpunt: 173…177 °C (1013 hPa) Dichtheid: 1,479 g/cm³ (20 °C) |
MDL-nummer:
MFCD00074940 CAS-nummer: 328-93-8 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...