p-Cymene

Leverancier: EHRENSTORFER GMBH
Gevaar

EHERCA14465500EA 70.7 EUR
EHERCA14465500
p-Cymene
p-Cymene
Formule: C₁₀H₁₄
MW: 134,22 g/mol
Kookpunt: 176 °C (1013 hPa)
Smeltpunt: –67,9 °C
Dichtheid: 0,8573 g/cm³ (20 °C)
Vlampunt: 47 °C
MDL-nummer: MFCD00008893
CAS-nummer: 99-87-6
EINECS: 202-796-7
UN: 2046
ADR: 3,III
Merck Index: 13,02792

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR