p-Cymene
Leverancier: Roth Carl
|
Gevaar
|
ROTH3465.1EA
67.9
EUR
ROTH3465.1
p-Cymene
p-Cymene
Formule:
C₁₀H₁₄ MW: 134,22 g/mol Kookpunt: 176 °C (1013 hPa) Smeltpunt: –67,9 °C Dichtheid: 0,8573 g/cm³ (20 °C) Vlampunt: 47 °C |
MDL-nummer:
MFCD00008893 CAS-nummer: 99-87-6 EINECS: 202-796-7 UN: 2046 ADR: 3,III Merck Index: 13,02792 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...