2-Fluorobiphenyl
Leverancier: EHRENSTORFER GMBH
EHERC13785000EA
147
EUR
EHERC13785000
2-Fluorobiphenyl
2-Fluorobiphenyl
Formule:
C₁₂H₉F MW: 172,2 g/mol Kookpunt: 248 °C (1013 hPa) Smeltpunt: 73…74 °C Dichtheid: 1,2452 g/cm³ (25 °C) |
MDL-nummer:
MFCD00000317 CAS-nummer: 321-60-8 EINECS: 206-290-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...