Bifenyl-4-ol
Leverancier: EHRENSTORFER GMBH
EHERC16070200EA
80
EUR
EHERC16070200
Bifenyl-4-ol
Bifenyl-4-ol
Formule:
C₁₂H₁₀O MW: 170,21 g/mol Kookpunt: 321 °C (1013 hPa) Smeltpunt: 162 °C Dichtheid: 1,29 g/cm³ (20 °C) |
MDL-nummer:
MFCD00002347 CAS-nummer: 92-69-3 EINECS: 202-179-2 Merck Index: 12,07459 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...