N,N-Dimethylcyclohexylamine
Leverancier: EHRENSTORFER GMBH
|
Gevaar
|
EHERC11830510EA
55.8
EUR
EHERC11830510
N,N-Dimethylcyclohexylamine
N,N-Dimethylcyclohexylamine
Formule:
C₈H₁₇N MW: 127,23 g/mol Kookpunt: 160…161 °C (1013 hPa) Smeltpunt: –60 °C Dichtheid: 0,849 g/cm³ (20 °C) Vlampunt: 41 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00003844 CAS-nummer: 98-94-2 EINECS: 202-715-5 UN: 2264 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...