Cyclopentylmethanol 98%
Leverancier: Thermo Scientific
ACRO147150100EA
127
EUR
ACRO147150100
Cyclopentylmethanol 98%
Cyclopentylmethanol
Formule:
C₆H₁₂O MW: 100,16 g/mol Kookpunt: 162 °C (1013 hPa) Dichtheid: 0,92 g/cm³ Vlampunt: 62 °C |
MDL-nummer:
MFCD00001384 CAS-nummer: 3637-61-4 EINECS: 222-861-3 |
Specificaties testresultaten
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4570 to 1.4590 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...