Potassium periodate ≥99.8% ACS, Fluka™
Leverancier: Honeywell Chemicals
|
Gevaar
|
210056-100GEA
104
EUR
210056-100G
Potassium periodate ≥99.8% ACS, Fluka™
Potassium periodate
Formule:
KlO₄ MW: 230 g/mol Smeltpunt: 582 °C Dichtheid: 3,62 g/cm³ (20 °C) Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00011407 CAS-nummer: 7790-21-8 EINECS: 232-196-0 UN: 3085 ADR: 5.1,I Merck Index: 12,07823 |
Specificaties testresultaten
Assay (excluding dried substance) | 99.8 - 100.3 % |
Manganese (Mn) | Max. 1 ppm |
Other halogens (as Cl) | Max. 0.01 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...