Magnesium fluoride, Patinal®
Leverancier: Merck
1.10261.1000EA
993
EUR
1.10261.1000
Magnesium fluoride, Patinal®
Magnesium fluoride
Formule:
MgF₂ MW: 62,3 g/mol Kookpunt: 2260 °C (1013 hPa) Smeltpunt: 1266 °C Dichtheid: 3 g/cm³ (20 °C) Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00011108 CAS-nummer: 7783-40-6 EINECS: 231-995-1 Merck Index: 12,05701 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...