Aluminium fluoride

Waarschuwing

HONE01277.9040
Aluminium fluoride
Aluminium fluoride
Formule: AlF₃
MW: 83,98 g/mol
Smeltpunt: 1040 °C
Dichtheid: 2,88 g/cm³ (20 °C)
MDL-nummer: MFCD00003426
CAS-nummer: 7784-18-1
EINECS: 232-051-1
UN: 3260
ADR: 8,II
Merck Index: 12,00351

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR