Lead(II) carbonate ≥99.999% (metals basis), Puratronic®
Leverancier: Thermo Scientific
|
Gevaar
|
10721.06EA
41.3
EUR
10721.06
10721.14
Lead(II) carbonate ≥99.999% (metals basis), Puratronic®
Lead(II) carbonate
Formule:
PbCO₃ MW: 267,21 g/mol Smeltpunt: 315 °C Dichtheid: 6,43 g/cm³ (20 °C) Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00064631 CAS-nummer: 598-63-0 EINECS: 209-943-4 UN: 2291 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...