Lead(II) carbonate, EMSURE® ACS for analysis, Supelco®
Leverancier: Merck
|
Gevaar
|
M-Clarity™ quality level = MQ300
1.07381.0250EA
111
EUR
1.07381.0250
1.07381.9999
Lead(II) carbonate, EMSURE® ACS for analysis, Supelco®
Lead(II) carbonate
Formule:
PbCO₃ MW: 267,21 g/mol Smeltpunt: 315 °C Dichtheid: 6,43 g/cm³ (20 °C) |
MDL-nummer:
MFCD00064631 CAS-nummer: 598-63-0 EINECS: 209-943-4 UN: 2291 ADR: 6.1,III |
Specificaties testresultaten
Solubility | (20 °C) practically insoluble |
Melting Point | 315 °C (decomposition) |
Molar Mass | 267.21 g/mol |
Bulk Density | 1300 kg/m3 |
Density | 6.6 g/cm3 (20 °C) |
pH | 5.8 (50 g/l, H2O, 20 °C) (slurry) |
Hazard Class | 6.1 |
Hazard Class Description | Toxic materials |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...