Octyltriethoxysilane for synthesis, Sigma-Aldrich®

Leverancier: Merck
Waarschuwing


M-Clarity™ quality level = MQ200

8.14414.0100EA 41.2 EUR
8.14414.0100 8.14414.9999
Octyltriethoxysilane for synthesis, Sigma-Aldrich®
Octyltriethoxysilane
Formule: C₁₄H₃₂O₃Si
MW: 276,49 g/mol
Kookpunt: 265 °C (1013 hPa)
Smeltpunt: –40 °C
Dichtheid: 0,8515 g/cm³ (20 °C)
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00039883
CAS-nummer: 2943-75-1
EINECS: 220-941-2

Bestel nu

Specificaties testresultaten

Solubility (20 °C) (decomposition)
Molar Mass 276.49 g/mol
Boiling Point 98 - 99 °C (2.7 hPa)
Vapor Pressure 0.1 hPa (20 °C)
Flash Point 109 °C
Refractive Index 1.415 (25 °C, 589 nm)
Density 0.87 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR