Tributyltin chloride analytical standard, Supelco®

Leverancier: Merck
Gevaar

442869.EA 140 EUR
442869.
Tributyltin chloride analytical standard, Supelco®
Tributyltin chloride
Formule: C₁₂H₂₇ClSn
MW: 325,51 g/mol
Kookpunt: 171…173 °C (25 mmHg)
Smeltpunt: –9 °C
Dichtheid: 1,2 g/cm³ (20 °C)
Vlampunt: >100 °C
MDL-nummer: MFCD00000521
CAS-nummer: 1461-22-9
EINECS: 215-958-7
UN: 2788
ADR: 6.1,II

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR