Chlorotriethylsilane
Leverancier: Molekula
MOLEM36065452EA
0
EUR
MOLEM36065452
Chlorotriethylsilane
Chlorotriethylsilane
Formule:
C₆H₁₅ClSi MW: 150,72 g/mol Kookpunt: 144 °C (1013 hPa) Dichtheid: 0,9 g/cm³ (20 °C) Vlampunt: 30 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00000507 CAS-nummer: 994-30-9 EINECS: 213-615-6 UN: 2986 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...