Tetramethylammonium chloride
Leverancier: Serva
|
Gevaar
|
Synoniemen:
TMACl
, TMA
SERA35924.01EA
0
EUR
SERA35924.01
Tetramethylammonium chloride
Tetramethylammonium chloride
Formule:
(CH₃)₄NCl MW: 109,60 g/mol Smeltpunt: > 300 °C Dichtheid: 1,169 g/cm³ (20 °C) Vlampunt: >100 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00011628 CAS-nummer: 75-57-0 EINECS: 200-880-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...