Tetrabutylammonium hydrogen sulphate, Supelco®

Leverancier: Merck
Waarschuwing

Synoniemen: tetra-n-butylammonium hexafluorophosphateTetrabutylammonium hydrogen sulfatetetrabutylammonium azide

M-Clarity™ quality level = MQ100

86853-10G-FEA 391 EUR
86853-10G-F 86853-50G-F
Tetrabutylammonium hydrogen sulphate, Supelco®
Tetrabutylammonium hydrogen sulphate
Formule: (C₄H₉)₄NHSO₄
MW: 339,54 g/mol
Smeltpunt: 169…172 °C
Dichtheid: 1,01 g/cm³ (25 °C)
MDL-nummer: MFCD00011637
CAS-nummer: 32503-27-8
EINECS: 251-068-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR