Tetraphenylphosphonium iodide for synthesis, Sigma-Aldrich®

Leverancier: Merck
Waarschuwing


M-Clarity™ quality level = MQ200

8.41105.0005EA 109 EUR
8.41105.0005
Tetraphenylphosphonium iodide for synthesis, Sigma-Aldrich®
Tetraphenylphosphonium iodide
Formule: C₂₄H₂₀IP
MW: 466,3 g/mol
Smeltpunt: 338…340 °C
Dichtheid: 1,5 g/cm³ (20 °C)
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00011917
CAS-nummer: 2065-67-0
EINECS: 218-179-0

Bestel nu

Specificaties testresultaten

Melting Point 338 - 340 °C
Molar Mass 466.3 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR