Diammoniumperoxodisulfaat (APS), Sigma-Aldrich®

Gevaar

Synoniemen: Ammonium persulphate

M-Clarity™ quality level = MQ200

SIAL09913-100GEA 46.4 EUR
SIAL09913-100G
Diammoniumperoxodisulfaat (APS), Sigma-Aldrich®
Diammoniumperoxodisulfaat (APS)
Formule: (NH₄)₂S₂O₈
MW: 228,2 g/mol
Smeltpunt: 120 °C
Dichtheid: 1,98 g/cm³ (20 °C)
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00003390
CAS-nummer: 7727-54-0
EINECS: 231-786-5
UN: 1444
ADR: 5.1,III
Merck Index: 13,00545
https://be.vwr.com/stibo/web/std.lang.all/06/39/16640639.jpg

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR