Ammonium sulphate ≥99.5%, Electran for molecular biology
Leverancier: VWR Chemicals
444445QEA 0 EUR
444445Q
Ammonium sulphate ≥99.5%, Electran for molecular biology
Ammonium sulphate
Formule: (NH₄)₂SO₄
MW: 132,14 g/mol
Smeltpunt: 336…339 °C
Dichtheid: 1,78 g/cm³ (20 °C)
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00003391
CAS-nummer: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559
Bestel nu
Specificaties testresultaten
Assay Min. 99.50 %
Appearance White crystalline powder
DNases (exo- and endonucleases) Not detected
RNases Not detected
Proteases Not detected
pH (5 %) 5.00 - 6.00
Absorbance (260 nm) (0.1 mol/l) Max. 0.01
Absorbance (280 nm) (0.1 mol/l) 0.01
NO3 (Nitrate) Max. 0.001 %
PO4 (Phosphate) Max. 0.0005 %
As (Arsenic) Max. 0.0001 %
Ca (Calcium) Max. 0.001 %
Cl (Chloride) Max. 0.001 %
Cu (Copper) Max. 0.0005 %
Fe (Iron) Max. 0.0005 %
K (Potassium) Max. 0.002 %
Mg (Magnesium) Max. 0.0005 %
Na (Sodium) Max. 0.002 %
Pb (Lead) Max. 0.0001 %
Zn (Zinc) Max. 0.0002 %