Ammonium sulphate, Millipore®
Leverancier: Merck Millipore Calbiochem
168355-1EA
273
EUR
168355-1
Ammonium sulphate, Millipore®
Ammonium sulphate
Formule:
(NH₄)₂SO₄ MW: 132,14 g/mol Smeltpunt: 336…339 °C Dichtheid: 1,78 g/cm³ (20 °C) Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00003391 CAS-nummer: 7783-20-2 EINECS: 231-984-1 REACH: 01-2119455044-46 Merck Index: 13,00559 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...