Dimethyl sulphite 99%
Leverancier: Thermo Scientific
|
Waarschuwing
|
ACRO148751000EA
161
EUR
ACRO148751000
Dimethyl sulphite 99%
Dimethyl sulphite
Formule:
C₂H₆O₃S MW: 110,13 g/mol Kookpunt: 126 °C (1013 hPa) Dichtheid: 1,29 g/cm³ Vlampunt: 30 °C |
MDL-nummer:
MFCD00008415 CAS-nummer: 616-42-2 EINECS: 210-481-0 UN: 1993 ADR: 3,III |
Specificaties testresultaten
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4085 to 1.4105 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...