Thiram

Leverancier: EHRENSTORFER GMBH
Waarschuwing

EHERC17570000EA 81.9 EUR
EHERC17570000
Thiram
Thiram
Formule: C₆H₁₂N₂S₄
MW: 240,44 g/mol
Kookpunt: 307 °C (1013 hPa)
Smeltpunt: 148…152 °C
Dichtheid: 1,426 g/cm³ (20 °C)
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00008325
CAS-nummer: 137-26-8
EINECS: 205-286-2
UN: 2811
ADR: 6.1,II
Merck Index: 12,09510

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR