Ninhydrin ACS
|
Waarschuwing
|
0544-100GEA
0
EUR
0544-100G
Ninhydrin ACS
Ninhydrin
Reacts with aldehydes and primary amines to form highly fluorescent ternary compounds.
Formule:
C₉H₆O₄ MW: 178,14 g/mol Kookpunt: 351 °C (1013 hPa) Smeltpunt: 250…258 °C Dichtheid: 0,862 g/cm³ (20 °C) Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00003791 CAS-nummer: 485-47-2 EINECS: 207-618-1 Merck Index: 12,06646 |
Specificaties testresultaten
Sensitivity to Amino Acids | Pass |
Solubility (1.0%, Water) | Pass |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...