Indole-5-carbonitrile

Leverancier: Molekula

MOLE17057160-5GEA 0 EUR
MOLE17057160-5G
Indole-5-carbonitrile
Indole-5-carbonitrile
Formule: C₉H₆N₂
MW: 142,16 g/mol
Smeltpunt: 106…108 °C
MDL-nummer: MFCD00005669
CAS-nummer: 15861-24-2
EINECS: 239-986-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR