Indole-5-carboxylic acid

Leverancier: Molekula

MOLEM26459684EA 0 EUR
MOLEM26459684
Indole-5-carboxylic acid
Indole-5-carboxylic acid
Formule: C₉H₇NO₂
MW: 161,16 g/mol
Smeltpunt: 211…213 °C
Opslagtemperatuur: Vriezer
MDL-nummer: MFCD00005678
CAS-nummer: 1670-81-1
EINECS: 216-799-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR