Indole-6-carboxylic acid ≥98%
|
Waarschuwing
|
L16019.03EA
55.2
EUR
L16019.03
Indole-6-carboxylic acid ≥98%
Indole-6-carboxylic acid
Formule:
C₉H₇NO₂ MW: 161,16 g/mol Smeltpunt: ∼ 258 °C (ontbinding) Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00210441 CAS-nummer: 1670-82-2 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...