5-Aminoindole

Leverancier: Molekula

MOLE53302534-1GEA 0 EUR
MOLE53302534-1G
5-Aminoindole
5-Aminoindole
Formule: C₈H₈N₂
MW: 132,16 g/mol
Kookpunt: 190 °C (6 mmHg)
Smeltpunt: 131…134 °C
MDL-nummer: MFCD00005679
CAS-nummer: 5192-03-0
EINECS: 225-977-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR