7-Ethylindole

Leverancier: Molekula

MOLE12743252-25GEA 0 EUR
MOLE12743252-25G
7-Ethylindole
7-Ethylindole
Formule: C₁₀H₁₁N
MW: 145,2 g/mol
Kookpunt: 230 °C (1013 hPa)
Dichtheid: 1,05 g/cm³ (20 °C)
MDL-nummer: MFCD00143514
CAS-nummer: 22867-74-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR