2-Phenylindole

Leverancier: Molekula

MOLEM67315784EA 0 EUR
MOLEM67315784
2-Phenylindole
2-Phenylindole
Formule: C₁₄H₁₁N
MW: 193,25 g/mol
Kookpunt: 420 °C (1013 hPa)
Smeltpunt: 186…187 °C
MDL-nummer: MFCD00005608
CAS-nummer: 948-65-2
EINECS: 213-436-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR