Neodymium trifluoride, Patinal®
Leverancier: Merck
1.06739.0100EA
0
EUR
1.06739.0100
Neodymium trifluoride, Patinal®
Neodymium trifluoride
Formule:
F₃Nd MW: 201,24 g/mol Kookpunt: 2327 °C (1013 hPa) Smeltpunt: 1410 °C Dichtheid: 6,5 g/cm³ (20 °C) Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00011135 CAS-nummer: 13709-42-7 EINECS: 237-253-3 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...