Cerium(IV) oxide ≥99.9%

Leverancier: Thermo Scientific
Waarschuwing

43818.09EA 65.4 EUR
43818.09 43818.18
Cerium(IV) oxide ≥99.9%
Cerium(IV) oxide

for vacuum deposition

Formule: CeO₂
MW: 172,11 g/mol
Kookpunt: 3468 °C (1013 hPa)
Smeltpunt: 2600 °C
Dichtheid: 7,132
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00010933
CAS-nummer: 1306-38-3
EINECS: 215-150-4
Merck Index: 12,02035

Bestel nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR