Ytterbium(III) oxide 99.99%

Leverancier: Thermo Scientific

ACRO264300100EA 124 EUR
ACRO264300100
Ytterbium(III) oxide 99.99%
Ytterbium(III) oxide
Formule: O₃Yb₂
MW: 394,08 g/mol
Smeltpunt: 2346 °C
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00011290
CAS-nummer: 1314-37-0
EINECS: 215-234-0

Bestel nu

Specificaties testresultaten

Appearance White Powder
Total trace metal impurities ≤100 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR