Koper(I)oxide ≥97% stabilised, powder <5 μm, Fluka™
|
Gevaar
|
208825-2KGEA
266
EUR
208825-2KG
Koper(I)oxide ≥97% stabilised, powder <5 μm, Fluka™
Koper(I)oxide
Formule:
Cu₂O MW: 143,09 g/mol Kookpunt: 1800 °C (1013 hPa) Smeltpunt: 1,230 °C Dichtheid: 6 g/cm³ (25 °C) Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00010974 CAS-nummer: 1317-39-1 EINECS: 215-270-7 ADR: 9,III Merck Index: 13,02694 |
Specificaties testresultaten
Contains | stabilizer |
Assay of Cu | 85.6 - 91.8 % |
Grain dissipation Horiba d50 | Max. 7.00 µm |
Form | powder |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...