Cadmiumfluoride ≥99.9%

Gevaar

36185.09EA 61.9 EUR
36185.09
Cadmiumfluoride ≥99.9%
Cadmiumfluoride
Formule: CdF₂
MW: 150,41 g/mol
Kookpunt: 1748 °C (1013 hPa)
Smeltpunt: 1065…1075 °C
Dichtheid: 6,33 g/cm³ (20 °C)
MDL-nummer: MFCD00010919
CAS-nummer: 7790-79-6
EINECS: 232-222-0
UN: 2570
ADR: 6.1,II
Merck Index: 12,01655

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR