Mercury(II) thiocyanate, EMSURE® Reag. Ph. Eur. for analysis, Supelco®

Gevaar


M-Clarity™ quality level = MQ300

1.04484.0100EA 178 EUR
1.04484.0100 1.04484.0025
Mercury(II) thiocyanate, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
Mercury(II) thiocyanate
Formule: C₂HgN₂S₂
MW: 316,76 g/mol
Smeltpunt: –38,9 °C
Dichtheid: 4 g/cm³ (20 °C)
Vlampunt: 120 °C
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00011039
CAS-nummer: 592-85-8
EINECS: 209-773-0
UN: 1646
ADR: 6.1,II
Merck Index: 12,05946

Bestel nu

Specificaties testresultaten

Assay (complexometric) ≥ 99.0 %
Fe (Iron) ≤ 0.002 %
Substances not reducing by formic acid (residue on ignition) ≤ 0.05 %
Corresponds to Reag. Ph Eur

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR