Niobium(V) oxide ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)

41081.KSEA 732 EUR
41081.KS
Niobium(V) oxide ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
Niobium(V) oxide
Formule: Nb₂O₅
MW: 265,81 g/mol
Smeltpunt: 1496 °C
Dichtheid: 4,6 g/cm³ (20 °C)
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00011128
CAS-nummer: 1313-96-8
EINECS: 215-213-6
Merck Index: 14,06559

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR