Dimidium bromide
Leverancier: Roth Carl
|
Waarschuwing
|
ROTH2217.1EA
112.5
EUR
ROTH2217.1
Dimidium bromide
Dimidium bromide
Formule:
C₂₀H₁₈BrN₃ MW: 380,29 g/mol Smeltpunt: 243…248 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00011757 CAS-nummer: 518-67-2 EINECS: 208-256-7 UN: 2811 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...