9-Methyladenine 98%
Leverancier: Thermo Scientific
|
Waarschuwing
|
ACRO325875000EA
414
EUR
ACRO325875000
ACRO325871000
9-Methyladenine 98%
9-Methyladenine
Formule:
C₆H₇N₅ MW: 149,16 g/mol Smeltpunt: 300…302 °C |
MDL-nummer:
MFCD00047232 CAS-nummer: 700-00-5 |
Specificaties testresultaten
Appearance | Beige Crystals |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...