1-Nitropyrene

Leverancier: EHRENSTORFER GMBH

EHERC20967100EA 118 EUR
EHERC20967100
1-Nitropyrene
1-Nitropyrene
Formule: C₁₆H₉NO₂
MW: 247,25 g/mol
Smeltpunt: 156 °C
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00004138
CAS-nummer: 5522-43-0
EINECS: 226-868-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR