5-Amino-2-chloropyridine
Leverancier: Molekula
|
Waarschuwing
|
MOLE85895134-1GEA
0
EUR
MOLE85895134-1G
5-Amino-2-chloropyridine
5-Amino-2-chloropyridine
Formule:
C₅H₅ClN₂ MW: 128,56 g/mol Kookpunt: 225 °C (1013 hPa) Smeltpunt: 83 °C |
MDL-nummer:
MFCD00006243 CAS-nummer: 5350-93-6 EINECS: 226-322-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...