4-Amino-2-chloropyridine
Leverancier: Molekula
|
Waarschuwing
|
MOLEM36211756EA
0
EUR
MOLEM36211756
4-Amino-2-chloropyridine
4-Amino-2-chloropyridine
Formule:
C₅H₅ClN₂ MW: 128,56 g/mol Kookpunt: 300 °C (1013 hPa) Smeltpunt: 87…89 °C |
MDL-nummer:
MFCD00060089 CAS-nummer: 14432-12-3 EINECS: 238-403-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...