Pyridine-2,5-dicarboxylic acid 98%

Leverancier: Thermo Scientific
Waarschuwing

ACRO131870050EA 25 EUR
ACRO131870050
Pyridine-2,5-dicarboxylic acid 98%
Pyridine-2,5-dicarboxylic acid
Formule: C₇H₅NO₄
MW: 167,12 g/mol
Kookpunt: 357 °C (1013 hPa)
Smeltpunt: 256 °C (ontbinding)
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00006297
CAS-nummer: 100-26-5
EINECS: 202-834-2
Merck Index: 13,05177

Bestel nu

Specificaties testresultaten

Appearance Yellow to green Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %
Sulfate (SO4) ≤0.005 %
Iron (Fe) ≤0.001 %
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.1 % (in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR