2-Amino-6-methyl-3-nitropyridine 98%

Waarschuwing

ACRO360780010EA 62.7 EUR
ACRO360780010
2-Amino-6-methyl-3-nitropyridine 98%
2-Amino-6-methyl-3-nitropyridine
Formule: C₆H₇N₃O₂
MW: 153,14 g/mol
Smeltpunt: 147…157 °C
MDL-nummer: MFCD00047443
CAS-nummer: 21901-29-1
UN: 2811

Bestel nu

Specificaties testresultaten

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 147 to 157 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR