4-Benzoylpyridine 98%
Leverancier: Thermo Scientific
|
Waarschuwing
|
ACRO154090050EA
28.3
EUR
ACRO154090050
4-Benzoylpyridine 98%
4-Benzoylpyridine
Formule:
C₁₂H₉NO MW: 183,21 g/mol Kookpunt: 315 °C (1013 hPa) Smeltpunt: 68…72 °C Vlampunt: 150 °C |
MDL-nummer:
MFCD00006430 CAS-nummer: 14548-46-0 EINECS: 238-586-7 |
Specificaties testresultaten
Appearance | Light yellow Crystalline powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 68 to 72 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...