Triphenylene 98%
Leverancier: Thermo Scientific
Gevaar
ACRO140360010EA 59.1 EUR
ACRO140360010 ACRO140360050
Triphenylene 98%
Triphenylene
Formule: C₁₈H₁₂
MW: 228,29 g/mol
Kookpunt: 438 °C (1013 hPa)
Smeltpunt: 196…200 °C
MDL-nummer: MFCD00001108
CAS-nummer: 217-59-4
EINECS: 205-922-9
UN: 3077
ADR: 9,III
Merck Index: 12,09870
Bestel nu
Specificaties testresultaten
Appearance White to beige Crystalline needles
Infrared spectrum Conforms
Melting point 196 to 200 °C
HPLC ≥97.5 %