Triphenylene

Leverancier: EHRENSTORFER GMBH
Gevaar

EHERC20945000EA 72.6 EUR
EHERC20945000
Triphenylene
Triphenylene
Formule: C₁₈H₁₂
MW: 228,29 g/mol
Kookpunt: 438 °C (1013 hPa)
Smeltpunt: 199 °C
Dichtheid: 1,302 g/cm³ (20 °C)
MDL-nummer: MFCD00001108
CAS-nummer: 217-59-4
EINECS: 205-922-9
UN: 3077
ADR: 9,III
Merck Index: 12,09870

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR