Bottle carrier
212-0702EA 78.8 EUR
VWRI212-0702
Bottle carrier
Made from 304 grade stainless steel, with a wire mesh bottom for increased stability.
  • Sturdy
  • Handles fold down for easier storage
  • Autoclavable
These safety wire bottle carriers will securely transport two ‘Merck’/Winchester style bottles up to 135 mm tall and 170 mm diameter (typically 2,5 litre capacity).