Agar
J10654.P2EA
113
EUR
J10654.P2
J10654.P5
J10654.Q1
Agar
Agar
Opslagtemperatuur:
Kamertemperatuur |
MDL-nummer:
MFCD00081288 CAS-nummer: 9002-18-0 EINECS: 232-658-1 |
Specificaties testresultaten
Total Ash | ≤6.5% |
Loss on Drying | ≤20.0% |
Identity | Passes Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...