Agar Noble agar, Ultrapure
Leverancier: Thermo Scientific
J10907.A1EA
407
EUR
J10907.A1
Agar Noble agar, Ultrapure
Agar
Gelling agent essentially free of impurities. Used mainly in biochemistry and molecular biology procedures wherever increased purity is required.
Smeltpunt:
90 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00081288 CAS-nummer: 9002-18-0 EINECS: 232-658-1 Merck Index: 13,00184 |
Specificaties testresultaten
Assay Absorbance (450 nm, 1.5%) | ≤0.150 |
Assay Gel Point (1.5%) | 32 - 37.5 °C |
Assay Gel Strength (1.5%) | ≥700 gm/cm² |
Assay Gel pH (1.5%) | 5.0 - 7.0 |
Ash | ≤1.6% |
Comment EEO (-Mr): | ≤0.450 (pH 8.4) |
Resistance (1% suspension) | ≥20,000 Ohms |
Residual water | ≤10% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...